missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl hydroperoxide, 70% Solution in water

Product Code. 10266090 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
250 g
500 g
2.5 kg
10 kg
Packaging:
Plastic Can
Plastic bottle
Unit Size:
10kg
2.5kg
250g
500g
5g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10266090 250 g Plastic bottle 250g
10703571 5 g Plastic bottle 5g
10347360 500 g Plastic bottle 500g
10649103 2.5 kg Plastic bottle 2.5kg
11914761 10 kg Plastic Can 10kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 10266090 Supplier Thermo Scientific Acros Supplier No. 180342500
ChemsChrom Mega Deals
Use promo code 27993
View all promotions
 Restricted Item
Estimated Shipment: 27-11-2026
Añadir a la cesta
Añadir a la cesta
Request Bulk or Custom Format

missing translation for 'validMainframeMsg'

Este artículo no se puede devolver. Vea la política de devoluciones

CAS: 75-91-2 | C4H10O2 | 90.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 75-91-2 , 7732-18-5
Molecular Formula C4H10O2
Molecular Weight (g/mol) 90.12
InChI Key CIHOLLKRGTVIJN-UHFFFAOYSA-N
Synonym tert-butyl hydroperoxide, tbhp, t-butyl hydroperoxide, tert-butylhydroperoxide, perbutyl h, t-butylhydroperoxide, cadox tbh, hydroperoxide, 1,1-dimethylethyl, 1,1-dimethylethyl hydroperoxide, terc. butylhydroperoxid
PubChem CID 6410
ChEBI CHEBI:64090
IUPAC Name 2-hydroperoxy-2-methylpropane
SMILES CC(C)(C)OO

Especificaciones

Melting Point -2.8°C
Color Colorless
Density 0.9400g/mL
Boiling Point 37.0°C (15.0 mmHg)
Flash Point 43°C
Packaging Plastic bottle
Linear Formula (CH3)3COOH
Refractive Index 1.3860 to 1.3880
Quantity 250 g
Beilstein 01,IV,1616
Fieser 01,88; 02,49; 03,37; 05,75; 06,81; 07,43; 08,62; 09,78; 10,64; 11,91; 12,88; 14,60; 15,55; 16,53; 17,56
Merck Index 15,1573
Specific Gravity 0.94
Solubility Information Solubility in water: miscible. Other solubilities: soluble in hydroxide solutions,soluble in most common organic solvents
Viscosity 4.1 mPa.s (20°C)
Formula Weight 90.12
Physical Form Liquid
Chemical Name or Material tert-Butyl hydroperoxide
Mostrar más Mostrar menos
compliance-icons compliance-icons compliance-icons compliance-icons compliance-icons
Nombre del producto
  • tert-Butyl hydroperoxide, 70% solution in water
Palabra de advertencia
  • Danger
Clasificación
  • Carcinogenicity Category 2
  • Skin corrosion/irritation Category 1 B
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Germ cell mutagenicity Category 2
  • Organic peroxide Type E & F
  • Skin sensitiser Category 1
Indicaciones de peligro
  • H317-May cause an allergic skin reaction.
  • H226-Flammable liquid and vapour.
  • H351-Suspected of causing cancer.
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H331-Toxic if inhaled.
  • H341-Suspected of causing genetic defects.
  • H411-Toxic to aquatic life with long lasting effects.
  • H311-Toxic in contact with skin.
  • H302-Harmful if swallowed.
  • H242-Heating may cause a fire.
Consejos de prudencia
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P220-Keep/Store away from clothing/combustible materials.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.