missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl isocyanide, 97%

Product Code. 10752392 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
Product Code. Quantity Packaging unitSize
10752392 1 g Glass bottle 1g
10229500 5 g Glass bottle 5g
2 options
This item is not returnable. View return policy

Product Code. 10752392

Brand: Thermo Scientific Acros 403600010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7188-38-7 | C5H9N | 83.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7188-38-7
Molecular Formula C5H9N
Molecular Weight (g/mol) 83.13
MDL Number MFCD00000002
InChI Key FAGLEPBREOXSAC-UHFFFAOYSA-N
Synonym tert-butyl isocyanide, tert-butylisocyanide, t-butylisonitrile, tert-butylisonitrile, tbin, propane, 2-isocyano-2-methyl, 2-methyl-2-isocyanopropane, 2-methylpropyl isocyanide, t-butyl isocyanide, 2-isocyano-2-methyl-propane
PubChem CID 23577
IUPAC Name 2-isocyano-2-methylpropane
SMILES CC(C)(C)[N+]#[C-]

Specifications

Density 0.7300g/mL
Boiling Point 91.0°C
Flash Point −2°C
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)3CNC
Refractive Index 1.3740 to 1.3780
Quantity 1 g
Fieser 02,50; 09,82; 11,99; 12,95; 14,63
Specific Gravity 0.73
Formula Weight 83.13
Percent Purity 97%
Chemical Name or Material tert-Butyl isocyanide, 97%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.