missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl isothiocyanate, 97%

Product Code. 11357785
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11357785 25 g 25g
11347785 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11357785 Supplier Thermo Scientific Alfa Aesar Supplier No. L02668.14
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 590-42-1 | C5H9NS | 115.19 g/mol

tert-Butyl isothiocyanate is a tool for the modification of the pore sizes on the top layer of heterostructured surface mounted metal-organic frameworks.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
tert-Butyl isothiocyanate is a tool for the modification of the pore sizes on the top layer of heterostructured surface mounted metal-organic frameworks.

Solubility
Reacts with water.

Notes
Moisture Sensitive. Store under dry inert gas. Incompatible with amines, alcohols and water.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 590-42-1
Molecular Formula C5H9NS
Molecular Weight (g/mol) 115.19
MDL Number MFCD00004816
InChI Key ZFWFRTVIIMTOLY-UHFFFAOYSA-N
Synonym tert-butyl isothiocyanate, t-butyl isothiocyanate, propane, 2-isothiocyanato-2-methyl, tert-butylisothiocyanate, tert.-butyl isothiocyanate, 2-isothiocyanato-2-methyl-propane, tert butyl isothiocyanate, tert-butyliso thiocyanate, butyl isothiocyanate, tert, t-butylisothiocyanate
PubChem CID 11544
IUPAC Name 2-isothiocyanato-2-methylpropane
SMILES CC(C)(C)N=C=S

Specifications

Melting Point 10°C to 12°C
Density 0.908
Boiling Point 141°C to 145°C
Flash Point 38°C (100°F)
Odor Pungent
Refractive Index 1.481
Quantity 25 g
UN Number UN2929
Beilstein 1361854
Sensitivity Moisture sensitive
Solubility Information Reacts with water.
Formula Weight 115.2
Percent Purity 97%
Chemical Name or Material tert-Butyl isothiocyanate
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • tert-Butyl isothiocyanate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H331-Toxic if inhaled.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: tert-Butyl isothiocyanate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.