missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl methyl ether, 99%

Product Code. 11348905
Change view
Click to view available options
Quantity:
500 mL
1000 mL
2500 mL
Unit Size:
1 L
2.5 L
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11348905 2500 mL 2.5 L
11328905 500 mL 500mL
11338905 1000 mL 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11348905 Supplier Thermo Scientific Alfa Aesar Supplier No. L14030.0F
 Restricted Item
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1634-04-4 | C5H12O | 88.15 g/mol

Used as a chromatography solvent. Acts as a fuel component in fuel for gasoline engines. They raise the oxygen content of gasoline(oxygenates).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as a chromatography solvent. Acts as a fuel component in fuel for gasoline engines. They raise the oxygen content of gasoline(oxygenates).

Solubility
Miscible with ethanol and ethyl ether. Immiscible with water.

Notes
Store in a cool, dry, well-ventilated area. It is hygroscopic in nature.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C5H12O
Molecular Weight (g/mol) 88.15
MDL Number MFCD00008812
InChI Key BZLVMXJERCGZMT-UHFFFAOYSA-N
Synonym tert-butyl methyl ether, methyl tert-butyl ether, mtbe, methyl t-butyl ether, t-butyl methyl ether, methyl tertiary-butyl ether, propane, 2-methoxy-2-methyl, methyl-tert-butyl ether, methyl-t-butyl ether, 2-methyl-2-methoxypropane
PubChem CID 15413
ChEBI CHEBI:27642
IUPAC Name 2-methoxy-2-methylpropane
SMILES CC(C)(C)OC

Specifications

Melting Point -109°C
Density 0.742
Quantity 2500 mL
Flash Point −25°C (−13°F)
Odor Mild
Refractive Index 1.369
UN Number UN2350
Merck Index 146032
Boiling Point 55°C to 56°C
Solubility Information Miscible with ethanol and ethyl ether. Immiscible with water.
Formula Weight 88.15
Percent Purity 0.99
Chemical Name or Material tert-Butyl methyl ether
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • tert-Butyl methyl ether
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H315-Causes skin irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
Supplemental information
  • MIXTURE LIST-Contain: Methyl tert-butyl ether

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.