missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl methyl ether, ACS reagent

Product Code. 11984701 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
Unit Size:
1 L
2.5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11984701 2.5 L 2.5 L
11974701 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11984701 Supplier Thermo Scientific Acros Supplier No. 447470025
  • ONLINE EXCLUSIVE PRICE
    102.00 €
    93.40 € / 2.5 L
    Save 8.60 €
    8% Off
View all promotions
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1634-04-4 | C5H12O | 88.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1634-04-4
Molecular Formula C5H12O
Molecular Weight (g/mol) 88.15
InChI Key BZLVMXJERCGZMT-UHFFFAOYSA-N
IUPAC Name 2-methoxy-2-methylpropane
SMILES COC(C)(C)C

Specifications

Melting Point -110.0°C
Density 0.7404 g/mL
Quantity 2.5 L
Flash Point -28°C
Packaging Glass Bottle
Refractive Index 1.3675 to 1.3695
Specific Gravity 0.7404
Linear Formula (CH3)3COCH3
Viscosity 0.36 mPa.s (20°C)
Merck Index 15, 6106
Boiling Point 54.0°C to 56.0°C
Solubility Information Solubility aq. soln.: 51 g/L (20°C). Other solubilities: miscible with most organic solvents, unstable in acid solutions, soly of water in methyl-t-butyl ether : 1,5 g/100 g
Formula Weight 88.15
Grade ACS
Chemical Name or Material tert-Butyl methyl ether
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • tert-Butyl methyl ether
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H315-Causes skin irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
Supplemental information
  • MIXTURE LIST-Contain: Methyl tert-butyl ether

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.