missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl methyl ether, CHROMASOLV™ Plus, for HPLC, Solstice

Product Code. 15666120
Change view
Click to view available options
Quantity:
1 L
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15666120 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15666120 Supplier Honeywell-Riedel-de Haen Supplier No. 6505601L
  • ONLINE EXCLUSIVE PRICE
    170.00 €
    146.42 € / 1 L
    Save 23.58 €
    13% Off
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CHROMASOLV™ Plus, for HPLC, 99.9%

  • For HPLC, spectrophotometry, and environmental analysis
  • Tested for HPLC-suitability via gradient analysis and UV-VIS spectroscopy
  • Low UV impurity profile
  • Low impurity gradient LC baseline
  • Very low levels of non-volatile impurities
TRUSTED_SUSTAINABILITY

Specifications

CAS 1634-04-4
Density 0.74g/mL at 25°C
Quantity 1 L
Explosion Limit 0.151
Flash Point −33°C (−27.4°F)
Ignition Point 705°F
Molecular Formula C5H12O
Linear Formula (CH3)3COCH3
MDL Number MFCD00008812
UN Number UN2398
Beilstein 1730942
Synonym tert-butyl methyl ether, methyl tert-butyl ether, mtbe, methyl t-butyl ether, t-butyl methyl ether, methyl tertiary-butyl ether, propane, 2-methoxy-2-methyl, methyl-tert-butyl ether, methyl-t-butyl ether, 2-methyl-2-methoxypropane
InChI Key BZLVMXJERCGZMT-UHFFFAOYSA-N
SMILES CC(C)(C)OC
IUPAC Name 2-methoxy-2-methylpropane
Molecular Weight (g/mol) 88.15
PubChem CID 15413
ChEBI CHEBI:27642
Formula Weight 88.15g/mol
Percent Purity 99%
Grade CHROMASOLV™ Plus
Chemical Name or Material tert-Butyl methyl ether
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • tert-Butyl methyl ether
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H315-Causes skin irritation.
Precautionary Statement
  • P243-Take precautionary measures against static discharge.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.