missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl methyl ether, For pesticide residue analysis, Honeywell Riedel-de Haën™

Product Code. 15654230
Change view
Click to view available options
Quantity:
1 L
2.5 L
4 x 2.5 L
Unit Size:
1 L
2.5 L
Pack of 4
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15654230 2.5 L 2.5 L
15644230 1 L 1 L
15664230 4 x 2.5 L Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15654230 Supplier Honeywell-Riedel-de Haen Supplier No. 344982.5L
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CHROMASOLV™, for pesticide residue analysis

  • For gas chromatography (GC)
  • Developed especially for the application in residue analysis of pesticides and other low-volatile substances using GC
  • Non-volatile matter and water content are specified for each solvent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1634-04-4
Molecular Formula C5H12O
Molecular Weight (g/mol) 88.15
MDL Number MFCD00008812
InChI Key BZLVMXJERCGZMT-UHFFFAOYSA-N
Synonym tert-butyl methyl ether, methyl tert-butyl ether, mtbe, methyl t-butyl ether, t-butyl methyl ether, methyl tertiary-butyl ether, propane, 2-methoxy-2-methyl, methyl-tert-butyl ether, methyl-t-butyl ether, 2-methyl-2-methoxypropane
PubChem CID 15413
ChEBI CHEBI:27642
IUPAC Name 2-methoxy-2-methylpropane
SMILES CC(C)(C)OC

Specifications

Density 0.74g/mL at 25°C
Quantity 2.5 L
Explosion Limit 0.151
Flash Point −33°C (−27.4°F)
Ignition Point 705°F
Linear Formula (CH3)3COCH3
UN Number UN2398
Beilstein 1730942
Formula Weight 88.15g/mol
Percent Purity ≥99.7%
Grade CHROMASOLV
Chemical Name or Material tert-Butyl methyl ether
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • tert-Butyl methyl ether
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H315-Causes skin irritation.
Precautionary Statement
  • P243-Take precautionary measures against static discharge.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : tert-butyl methyl ether
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.