missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl methyl ether, Honeywell™

Product Code. 16695803
Change view
Click to view available options
Quantity:
7 L
Unit Size:
7 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16695803 7 L 7 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16695803 Supplier Fluka Supplier No. 33020S7LRC
  • ONLINE EXCLUSIVE PRICE
    366.00 €
    300.77 € / 7 L
    Save 65.23 €
    17% Off
 Restricted Item
Estimated Shipment: 07-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 1634-04-4
CAS Max % 1
Melting Point -109°C
Color Colorless
Density 0.74 g/cm3 (20°C)
Boiling Point 55.3°C (1013 hPa)
Flash Point -28°C
Molecular Formula C5H12O
Quantity 7 L
InChI Key BZLVMXJERCGZMT-UHFFFAOYSA-N
SMILES COC(C)(C)C
IUPAC Name 2-methoxy-2-methylpropane
Molecular Weight (g/mol) 88.15
Chemical Name or Material tert-Butyl methyl ether
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.