missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl methyl ether, Puriss. p.a., ≥99.5% (GC), Solstice

Product Code. 15661740
Change view
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Glass Bottle
Unit Size:
1 L
2.5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15661740 1 L Glass Bottle 1 L
15671740 2.5 L Glass Bottle 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15661740 Supplier Honeywell-Riedel-de Haen Supplier No. 202561L
  • ONLINE EXCLUSIVE PRICE
    67.00 €
    62.93 € / 1 L
    Save 4.07 €
    6% Off
View all promotions
 Restricted Item
Estimated Shipment: 26-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 1634-04-4
Molecular Formula C5H12O
Molecular Weight (g/mol) 88.15
MDL Number MFCD00008812
InChI Key BZLVMXJERCGZMT-UHFFFAOYSA-N
Synonym tert-butyl methyl ether, methyl tert-butyl ether, mtbe, methyl t-butyl ether, t-butyl methyl ether, methyl tertiary-butyl ether, propane, 2-methoxy-2-methyl, methyl-tert-butyl ether, methyl-t-butyl ether, 2-methyl-2-methoxypropane
PubChem CID 15413
ChEBI CHEBI:27642
IUPAC Name 2-methoxy-2-methylpropane
SMILES CC(C)(C)OC

Specifications

Melting Point -109°C
Density 0.74g/mL at 25°C, 0.736 to 0.744g/mL at 20°C
Quantity 1 L
Explosion Limit 0.151
Flash Point −33°C (−27.4°F)
Ignition Point 705°F
Packaging Glass Bottle
Refractive Index 1.376
Linear Formula (CH3)3COCH3
UN Number UN2398
Beilstein 1730942
Boiling Point 55.3°C
Formula Weight 88.15g/mol
Percent Purity ≥99.5% (GC)
Grade puriss. p.a.
Chemical Name or Material Tert-Butyl Methyl Ether
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • tert-Butyl methyl ether
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H315-Causes skin irritation.
Precautionary Statement
  • P243-Take precautionary measures against static discharge.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.