missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl methyl sulfide, 98+%

Product Code. 11440507
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11440507 25 g 25g
11430507 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11440507 Supplier Thermo Scientific Alfa Aesar Supplier No. B22390.14
 Restricted Item
Estimated Shipment: 24-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 6163-64-0 | C5H12S | 104.21 g/mol

tert-Butyl methyl sulfide is used for LPG refining process and It have high content of sulfur.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
tert-Butyl methyl sulfide is used for LPG refining process and It have high content of sulfur.

Solubility
Difficult to mix.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6163-64-0
Molecular Formula C5H12S
Molecular Weight (g/mol) 104.21
MDL Number MFCD00008840
InChI Key CJFVCTVYZFTORU-UHFFFAOYSA-N
Synonym tert-butyl methyl sulfide, propane, 2-methyl-2-methylthio, sulfide, tert-butyl methyl, methyl tert-butyl sulfide, 3,3-dimethyl-2-thiabutane, 2-methyl-2-methylsulfanyl propane, t-butyl methyl sulfide, methyl t-butyl sulfide, tert-butyl methyl sulfane, acmc-1b7tx
PubChem CID 138679
IUPAC Name 2-methyl-2-methylsulfanylpropane
SMILES CSC(C)(C)C

Specifications

Density 0.826
Boiling Point 101°C to 102°C
Flash Point −3°C (26°F)
Odor Unpleasant
Linear Formula (CH3)3CSCH3
Refractive Index 1.44
Quantity 25 g
UN Number UN1993
Solubility Information Difficult to mix.
Formula Weight 104.22
Percent Purity ≥98%
Chemical Name or Material tert-Butyl methyl sulfide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.