missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl N-hydroxycarbamate, 98+%

Product Code. 10616154 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10616154 1 g Glass bottle 1g
10266500 5 g Glass bottle 5g
15129233 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10616154 Supplier Thermo Scientific Acros Supplier No. 209870010
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 36016-38-3 | C5H11NO3 | 133.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 36016-38-3
Molecular Formula C5H11NO3
Molecular Weight (g/mol) 133.15
MDL Number MFCD00002107
InChI Key DRDVJQOGFWAVLH-UHFFFAOYSA-N
Synonym n-boc-hydroxylamine, n-tert-butoxycarbonylhydroxylamine, tert-butyl hydroxycarbamate, boc-amino alcohol, carbamic acid, hydroxy-, 1,1-dimethylethyl ester, n-hydroxycarbamic acid tert-butyl ester, tert-butyl-n-hydroxycarbamate, n-tert-boc-hydroxylamine, t-butyl n-hydroxycarbamate, n-tert-butyloxycarbonylhydroxylamine
PubChem CID 97534
IUPAC Name tert-butyl N-hydroxycarbamate
SMILES CC(C)(C)OC(=O)NO

Specifications

Melting Point 53.0°C to 57.0°C
Color Pink to White
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Linear Formula HONHCO2C(CH3)3
Quantity 1 g
Beilstein 03,IV,168
Formula Weight 133.15
Percent Purity 98+%
Physical Form Crystalline Powder
Chemical Name or Material tert-Butyl N-hydroxycarbamate, 98+%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.