missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl nitrite, 90%, pure

Product Code. 11985161 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Plastic drum
Unit Size:
100g
2.5kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11985161 2.5 kg Plastic drum 2.5kg
10702071 100 g Glass bottle 100g
10452791 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.
Product Code. 11985161 Supplier Thermo Scientific Acros Supplier No. 216220025
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.

CAS: 540-80-7 | C4H9NO2 | 103.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 540-80-7
InChI Key IOGXOCVLYRDXLW-UHFFFAOYSA-N
Synonym t-butyl nitrite, 1,1-dimethylethyl nitrite, tert-butylnitrite, nitrous acid, 1,1-dimethylethyl ester, unii-4780h7u8lu, ccris 2098, alpha,alpha-dimethylethyl nitrite, nitrous acid, t-butyl ester, 2-methyl-2-propylnitrit, ch3 3cono
PubChem CID 10906
IUPAC Name tert-butyl nitrite
SMILES CC(C)(C)ON=O

Specifications

Color Yellow
Density 0.8600g/mL
Boiling Point 61.0°C to 63.0°C
Flash Point −10°C
Infrared Spectrum Authentic
Assay Percent Range 88% min. (GC)
Packaging Plastic drum
Molecular Formula C4H9NO2
Linear Formula (CH3)3CNO2
Refractive Index 1.3670 to 1.3700
Quantity 2.5 kg
Beilstein 01,382
Fieser 07,48
Merck Index 15,1586
Specific Gravity 0.86
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in alcohol,ether,chloroform and carbon,disulfide,practically insoluble in glycerol
Molecular Weight (g/mol) 103.12
Formula Weight 103.12
Percent Purity 90%
Grade Pure
Physical Form Liquid
Chemical Name or Material tert-Butyl nitrite
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • tert-Butyl nitrite
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Flammable liquid Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.