missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl Phenyl Carbonate, TRC

Product Code. 30331458
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30331458 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30331458 Supplier Toronto Research Chemicals Supplier No. TRCB8093005G
 Restricted Item
Estimated Shipment: 01-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material tert-Butyl Phenyl Carbonate
CAS 6627-89-0
Molecular Formula C11H14O3
Quantity 5 g
Synonym Carbonic Acid tert-Butyl Phenyl Ester, 1,1-Dimethylethyl Phenyl Carbonate, 1-tert-Butoxycarbonyloxybenzene, NSC 60248, Phenyl tert-Butyl Carbonate
InChI Formula InChI=1S/C11H14O3/c1-11(2,3)14-10(12)13-9-7-5-4-6-8-9/h4-8H,1-3H3
SMILES CC(C)(C)OC(=O)Oc1ccccc1
IUPAC Name tert-butyl phenyl carbonate
Molecular Weight (g/mol) 194.23
Formula Weight 194.0943
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.