missing translation for 'onlineSavingsMsg'
Learn More

tert-butyl (S)-4-(dimethylamino)-1-(piperazine-1-carbonyl)butylcarbamate, Thermo Scientific™

Product Code. 10587770
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10587770 250 mg Amber Glass Bottle 250mg
10301380 1 g Amber Glass Bottle 1g
10687390 5 g Amber Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10587770 Supplier Thermo Scientific Maybridge Supplier No. MO08535CB
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 1174064-68-6
Molecular Formula C16H32N4O3
Molecular Weight (g/mol) 328.457
InChI Key CDZDEHVWAXDQAQ-ZDUSSCGKSA-N
Synonym tert-butyl s-4-dimethylamino-1-piperazine-1-carbonyl butylcarbamate, tert-butyl n-2s-5-dimethylamino-1-oxo-1-piperazin-1-yl pentan-2-yl carbamate, tert-butyl 2s-5-dimethylamino-1-oxo-1-piperazin-1-yl pentan-2-yl carbamate
PubChem CID 45594327
IUPAC Name tert-butyl N-[(2S)-5-(dimethylamino)-1-oxo-1-piperazin-1-ylpentan-2-yl]carbamate
SMILES CC(C)(C)OC(=O)NC(CCCN(C)C)C(=O)N1CCNCC1

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 328.45
Percent Purity 97%
Chemical Name or Material tert-Butyl (S)-4-(dimethylamino)-1-(piperazine-1-carbonyl)butylcarbamate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.