missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl (S)-(-)-4-formyl-2,2-dimethyl-3-oxazolidinecarboxylate, 95%, Thermo Scientific Chemicals

Product Code. 10290453 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10290453 250 mg 250mg
10404442 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10290453 Supplier Thermo Scientific Acros Supplier No. 386572500
 Restricted Item
Estimated Shipment: 19-08-2026
Add to basket
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 102308-32-7
Molecular Formula C11H19NO4
Molecular Weight (g/mol) 229.28
MDL Number MFCD00209557
InChI Key PNJXYVJNOCLJLJ-MRVPVSSYSA-N
Synonym s---3-boc-2,2-dimethyloxazolidine-4-carboxaldehyde, tert-butyl s---4-formyl-2,2-dimethyl-3-oxazolidinecarboxylate, tert-butyl 4s-4-formyl-2,2-dimethyl-1,3-oxazolidine-3-carboxylate, unii-41aj913hqi, s-tert-butyl 4-formyl-2,2-dimethyloxazolidine-3-carboxylate, 1,1-dimethylethyl-s-4-formyl-2,2-dimethyl-3-oxazolidine carboxylate, s---3-tert-butoxycarbonyl-4-formyl-2,2-dimethyl-1,3-oxazolidine, s-4-formyl-2,2-dimethyl-oxazolidine-3-carboxylic acid tert-butyl ester, 3-oxazolidinecarboxylic acid, 4-formyl-2,2-dimethyl-, 1,1-dimethylethyl ester, 4s, s---3-boc-4-formyl-2,2-dimethyl-1,3-oxazolidine
PubChem CID 179824
IUPAC Name tert-butyl (4S)-4-formyl-2,2-dimethyl-1,3-oxazolidine-3-carboxylate
SMILES CC1(N(C(CO1)C=O)C(=O)OC(C)(C)C)C

Specifications

Color Colorless to Yellow
Density 1.0600g/mL
Boiling Point 83°C to 88°C (1.0 mmHg)
Flash Point 108°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass Bottle
Refractive Index 1.4450 to 1.4490 (20°C, 589nm)
Quantity 250 mg
Merck Index 15,4404
Specific Gravity 1.06
Specific Rotation -85° to -95° (20°C, 589nm) (c=1, CHCl3)
Formula Weight 229.28
Percent Purity 95%
Physical Form Oil
Chemical Name or Material tert-Butyl (S)-(-)-4-formyl-2, 2-dimethyl-3-oxazolidinecarboxylate
Show More Show Less
compliance-icons
Product Identifier
  • tert-Butyl (S)-(-)-4-formyl-2,2-dimethyl-3-oxazolidinecarboxylate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.