missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl trimethylacetate, 98%, Thermo Scientific™

Product Code. 10637854 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
:
1g; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10637854 1g; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10637854 Supplier Thermo Scientific Acros Supplier No. 387620010
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 16474-43-4
Density 0.8130g/mL
Boiling Point 134.0°C to 135.0°C
Flash Point >62°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Molecular Formula C9H18O2
Linear Formula (CH3)3CCO2C(CH3)3
Refractive Index 1.3910 to 1.3930
MDL Number MFCD01861974
Quantity 1g
Specific Gravity 0.813
Synonym tert-butyl pivalate, tert-butyl trimethylacetate, propanoic acid, 2,2-dimethyl, 1,1-dimethylethyl ester, di-tert-butylcarboxylic acid, 2,2-dimethylpropanoic acid tert-butyl ester, 2,2-dimethyl-propionic acid tert-butyl ester, propanoic acid, 2,2-dimethyl-, 1,1-dimethylethyl ester, propanoic acid,2,2-dimethyl-, 1,1-dimethylethyl ester
InChI Key VXHFNALHLRWIIU-UHFFFAOYSA-N
SMILES CC(C)(C)OC(=O)C(C)(C)C
IUPAC Name tert-butyl 2,2-dimethylpropanoate
Molecular Weight (g/mol) 158.24
PubChem CID 519272
Formula Weight 158.24
Percent Purity 98%
Chemical Name or Material tert-Butyl trimethylacetate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.