missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl vinyl ether, 98%, stabilized

Product Code. 10341693 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
10 mL
50 mL
Packaging:
Glass bottle
Unit Size:
10mL
50mL
This item is not returnable. View return policy

Product Code. 10341693

Brand: Thermo Scientific Acros 379120100

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 926-02-3 | C6H12O | 100.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 926-02-3
Molecular Formula C6H12O
Molecular Weight (g/mol) 100.16
MDL Number MFCD00048246
InChI Key PGYJSURPYAAOMM-UHFFFAOYSA-N
Synonym tert-butyl vinyl ether, 2-ethenyloxy-2-methylpropane, propane, 2-ethenyloxy-2-methyl, propane,2-ethenyloxy-2-methyl, t-butyl vinyl ether, tert-butylvinyl ether, vinyltertiary-butylether, tertiarybutyl vinyl ether, vinyl tert.-butyl ether, tertiary butyl vinyl ether
PubChem CID 70220
IUPAC Name 2-ethenoxy-2-methylpropane
SMILES CC(C)(C)OC=C

Specifications

Color Colorless to Yellow
Density 0.7620g/mL
Boiling Point 78°C
Flash Point -17°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.3970 to 1.399
Quantity 10 mL
Specific Gravity 0.762
Solubility Information Solubility in water: sparingly soluble
Stabilizer approx. 100ppm KOH
Viscosity 0.41 mPa.s (20°C)
Formula Weight 100.16
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material tert-Butyl vinyl ether
Show More Show Less
compliance-icons
Product Identifier
  • tert-Butyl vinyl ether
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Flammable liquid Category 2
Hazard Statement
  • H412-Harmful to aquatic life with long lasting effects.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P273-Avoid release to the environment.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.