missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Beskrivning
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifikationer
Specifikationer
| Color | Colorless to Yellow |
| Density | 0.7620g/mL |
| Boiling Point | 78°C |
| Flash Point | -17°C |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 97.5% min. (GC) |
| Packaging | Glass bottle |
| Refractive Index | 1.3970 to 1.399 |
| Quantity | 50 mL |
| Specific Gravity | 0.762 |
| Visa mer |
Säkerhet och hantering
Produktbeteckning
- tert-Butyl vinyl ether
- Danger
- LONG-TERM AQUATIC HAZARD Chronic 3
- Flammable liquid Category 2
- H412-Harmful to aquatic life with long lasting effects.
- H225-Highly flammable liquid and vapour.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P240-Ground/bond container and receiving equipment.
- P273-Avoid release to the environment.
RUO – Research Use Only
Produkttitel
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.
Hittar du en möjlighet till förbättring?