missing translation for 'onlineSavingsMsg'
Learn More

tert-Butylacetic Acid, 98%

Product Code. 10346120 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 mL
50 mL
250 mL
Packaging:
Glass bottle
Unit Size:
10mL
250mL
50mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10346120 10 mL Glass bottle 10mL
10204710 50 mL Glass bottle 50mL
10519823 250 mL Glass bottle 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10346120 Supplier Thermo Scientific Acros Supplier No. 107770100
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1070-83-3 | C6H12O2 | 116.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1070-83-3
Molecular Formula C6H12O2
Molecular Weight (g/mol) 116.16
MDL Number MFCD00002715
InChI Key MLMQPDHYNJCQAO-UHFFFAOYSA-N
Synonym 3,3-dimethylbutyric acid, tert-butylacetic acid, t-butylacetic acid, butanoic acid, 3,3-dimethyl, 3,3-dimethyl-butanoic acid, unii-q80k0tzq55, butyric acid, 3,3-dimethyl, 3,3-dimethyl-butyric acid, 3,3-dimethyl-n-butyric acid, tert-butyric acid
PubChem CID 14057
ChEBI CHEBI:38647
IUPAC Name 3,3-dimethylbutanoic acid
SMILES CC(C)(C)CC(=O)O

Specifications

Melting Point -8°C to 6°C
Density 0.9100g/mL
Boiling Point 185.0°C to 190.0°C
Flash Point 88°C
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula (CH3)3CCH2CO2H
Refractive Index 1.4090 to 1.4110
Quantity 10 mL
Beilstein 02, 337
Merck Index 15, 1540
Specific Gravity 0.91
Solubility Information Solubility in water: 19g/L (20 c)
Formula Weight 116.16
Percent Purity 98%
Chemical Name or Material tert-Butylacetic acid
Show More Show Less
compliance-icons
Product Identifier
  • tert-Butylacetic acid
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.