missing translation for 'onlineSavingsMsg'
Learn More

tert-Butylacetyl chloride, 98+%

Product Code. 11453950
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Unit Size:
100g
25g
500g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11453950 5 g 5g
11459442 25 g 25g
11469442 100 g 100g
11479442 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11453950 Supplier Thermo Scientific Alfa Aesar Supplier No. A10162.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7065-46-5 | C6H11ClO | 134.603 g/mol

tert-Butylacetyl chloride is used an acyl chloride compound for proteomics research. It is also used as intermediate for important acylation reagents agrochemical and pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
tert-Butylacetyl chloride is used an acyl chloride compound for proteomics research. It is also used as intermediate for important acylation reagents agrochemical and pharmaceutical.

Solubility
Reacts with water.

Notes
Moisture Sensitive Store away from strong oxidizing agents. keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7065-46-5
Molecular Formula C6H11ClO
Molecular Weight (g/mol) 134.603
MDL Number MFCD00000737
InChI Key BUTKIHRNYUEGKB-UHFFFAOYSA-N
Synonym 3,3-dimethylbutyryl chloride, tert-butylacetyl chloride, t-butylacetyl chloride, butanoyl chloride, 3,3-dimethyl, 3,3-dimethyl butyryl chloride, tert-butyl acetyl chloride, neohexanoyl chloride, t-butylacetylchloride, tertbutylacetylchloride, pubchem2093
PubChem CID 81514
IUPAC Name 3,3-dimethylbutanoyl chloride
SMILES CC(C)(C)CC(=O)Cl

Specifications

Boiling Point 128°C to 130°C
Flash Point 21°C (69°F)
Odor Acrid
Quantity 5 g
UN Number UN2924
Beilstein 1740421
Formula Weight 134.61
Percent Purity ≥98%
Chemical Name or Material tert-Butylacetyl chloride

RUO – Research Use Only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.