missing translation for 'onlineSavingsMsg'
Learn More

tert-Butylacetyl chloride, 99%

Product Code. 10094093 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 mL
50 mL
250 mL
Packaging:
Glass bottle
Unit Size:
10mL
250mL
50mL
Product Code. Quantity Packaging unitSize
10094093 10 mL Glass bottle 10mL
10705751 50 mL Glass bottle 50mL
10599990 250 mL Glass bottle 250mL
3 options
This item is not returnable. View return policy

Product Code. 10094093

Brand: Thermo Scientific Acros 107790100

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7065-46-5 | C6H11ClO | 134.61 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7065-46-5
Molecular Formula C6H11ClO
Molecular Weight (g/mol) 134.61
MDL Number MFCD00000737
InChI Key BUTKIHRNYUEGKB-UHFFFAOYSA-N
Synonym 3,3-dimethylbutyryl chloride, tert-butylacetyl chloride, t-butylacetyl chloride, butanoyl chloride, 3,3-dimethyl, 3,3-dimethyl butyryl chloride, tert-butyl acetyl chloride, neohexanoyl chloride, t-butylacetylchloride, tertbutylacetylchloride, pubchem2093
PubChem CID 81514
IUPAC Name 3,3-dimethylbutanoyl chloride
SMILES CC(C)(C)CC(=O)Cl

Specifications

Color Colorless to Yellow
Density 0.9600g/mL
Boiling Point 127.0°C to 129.0°C
Flash Point 21°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula (CH3)3CCH2COCl
Refractive Index 1.4210 to 1.4230
Quantity 10 mL
Specific Gravity 0.96
Solubility Information Solubility in water: reacts. Other solubilities: soluble in ethanol
Formula Weight 134.61
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material tert-Butylacetyl chloride, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.