missing translation for 'onlineSavingsMsg'
Learn More

tert-Butylamine, 98%

Product Code. 11469033
Change view
Click to view available options
Quantity:
10,000 mL
2500 mL
500 mL
Unit Size:
10000mL
2500mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11469033 500 mL 500mL
11479033 2500 mL 2500mL
11489033 10,000 mL 10000mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
Product Code. 11469033 Supplier Thermo Scientific Alfa Aesar Supplier No. A11135.AP
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

CAS: 75-64-9 | C4H11N | 73.139 g/mol

tert-butylamine is used as a precursor for the preparation of rubber accelerators, pharmaceuticals and dyes. It is employed in synthesis of N-tert-butyl-acetamide, isoquinolines, pyridines, decumbenine, sulfontes, sulfonamides and methyl beta-ketophosphonate. It is also used in the study the effect of amines in influencing atmospheric sulfuric acid - water nucleation, hydroamination of terminal alkynes and measuring the surface acidity of solid catalyst. It acts as an indicator molecule in the study of NMR -based pH determination method.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
tert-butylamine is used as a precursor for the preparation of rubber accelerators, pharmaceuticals and dyes. It is employed in synthesis of N-tert-butyl-acetamide, isoquinolines, pyridines, decumbenine, sulfontes, sulfonamides and methyl beta-ketophosphonate. It is also used in the study the effect of amines in influencing atmospheric sulfuric acid - water nucleation, hydroamination of terminal alkynes and measuring the surface acidity of solid catalyst. It acts as an indicator molecule in the study of NMR -based pH determination method.

Solubility
Miscible with water, alcohol, ether, chloroform and organic solvents.

Notes
Air sensitive. Incompatible with strong oxidizing agents, strong acids and acid anhydrides.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-64-9
Molecular Formula C4H11N
MDL Number MFCD00008050
InChI Key YBRBMKDOPFTVDT-UHFFFAOYSA-N
Synonym tert-butylamine, erbumine, 2-amino-2-methylpropane, t-butylamine, 2-propanamine, 2-methyl, trimethylaminomethane, 1,1-dimethylethylamine, 2-aminoisobutane, 1,1-dimethylethanamine, butylamine, tert
PubChem CID 6385
ChEBI CHEBI:44639
IUPAC Name 2-methylpropan-2-amine
SMILES CC(C)(C)N

Specifications

Melting Point -67°C
Density 0.696
Boiling Point 44°C to 46°C
Flash Point −38°C (−36°F)
Odor Amine-like
Refractive Index 1.378
Quantity 500 mL
UN Number UN2733
Beilstein 605267
Sensitivity Air Sensitive
Merck Index 14,1545
Solubility Information Miscible with water,alcohol,ether,chloroform and organic solvents.
Molecular Weight (g/mol) 73.139
Formula Weight 73.14
Percent Purity 98%
Chemical Name or Material tert-Butylamine
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • tert-Butylamine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H331-Toxic if inhaled.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: tert-Butylamine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.