missing translation for 'onlineSavingsMsg'
Learn More

tert-Butylchlorodimethylsilane, 1.0M solution in dichloromethane, AcroSeal™

Product Code. 10661555 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
Packaging:
AcroSeal™ Glass bottle
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10661555 100 mL AcroSeal™ Glass bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10661555 Supplier Thermo Scientific Acros Supplier No. 369101000
 Restricted Item
Estimated Shipment: 22-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 18162-48-6 | C6H15ClSi | 150.72 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 18162-48-6, 75-09-2
Color Colorless
Density 1.2250g/mL
Flash Point 58°C
Packaging AcroSeal™ Glass bottle
Molecular Formula C6H15ClSi
MDL Number MFCD00000501
Quantity 100 mL
Merck Index 15, 1567
Specific Gravity 1.225
Synonym tert-butyldimethylsilyl chloride, tert-butyldimethylchlorosilane, t-butyldimethylchlorosilane, tert-butylchlorodimethylsilane, tbdms chloride, tert-butyl chloro dimethylsilane, chloro-tert-butyldimethylsilane, silane, chloro 1,1-dimethylethyl dimethyl, tbscl, t-butyldimethylsilyl chloride
InChI Key BCNZYOJHNLTNEZ-UHFFFAOYSA-N
SMILES CC(C)(C)[Si](C)(C)Cl
IUPAC Name tert-butyl-chloro-dimethylsilane
Molecular Weight (g/mol) 150.72
PubChem CID 28928
ChEBI CHEBI:85071
Formula Weight 150.72
Physical Form Liquid
Chemical Name or Material tert-Butylchlorodimethylsilane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.