missing translation for 'onlineSavingsMsg'
Learn More

tert-Butylcyclohexane, 99+%

Product Code. 11431137
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
Product Code. Quantity unitSize
11431137 100 g 100g
11421137 25 g 25g
2 options
This item is not returnable. View return policy

Product Code. 11431137

Brand: Thermo Scientific Alfa Aesar B24126.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3178-22-1 | C10H20 | 140.27 g/mol

Used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3178-22-1
Molecular Formula C10H20
Molecular Weight (g/mol) 140.27
MDL Number MFCD00001471
InChI Key XTVMZZBLCLWBPM-UHFFFAOYSA-N
Synonym t-butylcyclohexane, cyclohexane, tert-butyl, 1-tert-butylcyclohexane, cyclohexane, 1,1-dimethylethyl, unii-0rgt19ucq6, 1,1-dimethylethyl cyclohexane, 0rgt19ucq6, t-butyl cyclohexane, 4-t-butylcyclohexane, tert-butyl-cyclohexane
PubChem CID 18508
IUPAC Name tert-butylcyclohexane
SMILES CC(C)(C)C1CCCCC1

Specifications

Melting Point -41°C
Density 0.83
Boiling Point 171°C
Flash Point 40°C (104°F)
Refractive Index 1.447
Quantity 100 g
UN Number UN3295
Solubility Information Insoluble in water.
Formula Weight 140.27
Percent Purity ≥99%
Chemical Name or Material tert-Butylcyclohexane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.