missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyldichlorophosphine, Thermo Scientific Chemicals

Product Code. 10404735 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
2.5 g
10 g
Packaging:
Glass bottle
Unit Size:
10g
2.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10404735 10 g Glass bottle 10g
10195790 2.5 g Glass bottle 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.
Product Code. 10404735 Supplier Thermo Scientific Acros Supplier No. 300530100
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.

CAS: 25979-07-1 | Molecular Formula: C4H9Cl2P | Molecular Weight: 158.98 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25979-07-1
Molecular Formula C4H9Cl2P
Molecular Weight (g/mol) 158.98
MDL Number MFCD00013615
InChI Key NMJASRUOIRRDSX-UHFFFAOYSA-N
Synonym tert-butyldichlorophosphine, t-butyldichlorophosphine, tert-c4h9pcl2, tert-butyl dichloro phosphane, phosphonous dichloride, 1,1-dimethylethyl, snpladsptpdwuup@, pubchem6471, acmc-1cmec, tert-butyl dichloro phosphine, tert-butylphosphine dichloride
PubChem CID 117690
IUPAC Name tert-butyl(dichloro)phosphane
SMILES CC(C)(C)P(Cl)Cl

Specifications

Melting Point 49.0°C
Color White to Yellow
Boiling Point 142.0°C to 150.0°C
Infrared Spectrum Authentic
Packaging Glass bottle
Linear Formula (CH3)3CPCl2
Quantity 10 g
Solubility Information Solubility in water: reacts. Other solubilities: soluble in n-pentane,n-hexane,ether,toluene,,thf
Formula Weight 158.98
Physical Form Crystalline Low Melting Solid
Chemical Name or Material tert-Butyldichlorophosphine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.