missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyldimethylsilyl chloride, 50 wt.% solution in toluene, AcroSeal™

Product Code. 10067949 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10067949 100 mL AcroSeal™ Glass Bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10067949 Supplier Thermo Scientific Acros Supplier No. 433111000
  • ONLINE EXCLUSIVE PRICE
    108.00 €
    100.00 € / 100mL
    Save 8.00 €
    7% Off
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 18162-48-6 | C6H15ClSi | 150.72 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material tert-Butyldimethylsilyl chloride
CAS 18162-48-6, 108-88-3
Color Colorless
Density 0.8660g/mL
Boiling Point 110.0°C to 126.0°C
Flash Point 4°C
Packaging AcroSeal™ Glass Bottle
Molecular Formula C6H15ClSi
Quantity 100 mL
Merck Index 15,1567
Specific Gravity 0.866
Synonym tert-butyldimethylsilyl chloride, tert-butyldimethylchlorosilane, t-butyldimethylchlorosilane, tert-butylchlorodimethylsilane, tbdms chloride, tert-butyl chloro dimethylsilane, chloro-tert-butyldimethylsilane, silane, chloro 1,1-dimethylethyl dimethyl, tbscl, t-butyldimethylsilyl chloride
InChI Key BCNZYOJHNLTNEZ-UHFFFAOYSA-N
SMILES CC(C)(C)[Si](C)(C)Cl
IUPAC Name tert-butyl-chloro-dimethylsilane
Molecular Weight (g/mol) 150.72
PubChem CID 28928
ChEBI CHEBI:85071
Formula Weight 150.72
Percent Purity 48 to 52%
Physical Form Liquid
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • tert-Butyldimethylsilyl chloride, 50 wt.% solution in toluene
Signal Word
  • Danger
Hazard Category
  • Aspiration toxicity Category 1
  • Flammable liquid Category 2
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
  • H314-Causes severe skin burns and eye damage.
  • H361d-Suspected of damaging the unborn child.
  • H373-May cause damage to organs.
  • H336-May cause drowsiness or dizziness.
  • H225-Highly flammable liquid and vapour.
  • EUH014-Reacts violently with water.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P402+P404-Store in a dry place. Store in a closed container.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.