missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyldimethylsilyl trifluoromethanesulfonate, 98%

Product Code. 10012760 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
Packaging:
Glass bottle
Unit Size:
25mL
5mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10012760 5 mL Glass bottle 5mL
11376175 25 mL Glass bottle 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10012760 Supplier Thermo Scientific Acros Supplier No. 209860050
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 69739-34-0 | C7H15F3O3SSi | 264.33 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 69739-34-0
Molecular Formula C7H15F3O3SSi
Molecular Weight (g/mol) 264.33
MDL Number MFCD00000405
InChI Key WLLIXJBWWFGEHT-UHFFFAOYSA-N
Synonym tert-butyldimethylsilyl trifluoromethanesulfonate, trifluoromethanesulfonic acid tert-butyldimethylsilyl ester, tbdms triflate, t-butyldimethylsilyl triflate, tert-butyldimethylsilyl trifluoromethanesulphonate, tbdms-otf, tert-butyldimethylsilyl triflate, silane tbm2 triflate, t-butyldimethylsilyltrifluoromethanesulfonate, t-butylmethylsilyl triflate
PubChem CID 123564
IUPAC Name [tert-butyl(dimethyl)silyl] trifluoromethanesulfonate
SMILES CC(C)(C)[Si](C)(C)OS(=O)(=O)C(F)(F)F

Specifications

Color Colorless to Yellow
Density 1.1500g/mL
Boiling Point 65°C to 67°C (12.0 mmHg)
Flash Point 36°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min
Packaging Glass bottle
Linear Formula CF3SO3Si(CH3)2C(CH3)3
Refractive Index 1.3850 to 1.388
Quantity 5 mL
Fieser 10,63; 11,90; 12,86; 13,50; 17,55
Specific Gravity 1.15
Solubility Information Solubility in water: decomposes
Formula Weight 264.33
Percent Purity 98%
Physical Form Fuming Liquid
Chemical Name or Material tert-Butyldimethylsilyl trifluoromethanesulfonate
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • tert-Butyldimethylsilyl trifluoromethanesulfonate
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 3
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.