missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyldimethylsilyl Trifluoromethanesulfonate, TRC

Product Code. 30329536
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30329536 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30329536 Supplier Toronto Research Chemicals Supplier No. TRCB69177510G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material tert-Butyldimethylsilyl triflate
CAS 69739-34-0
Molecular Formula C7 H15 F3 O3 S Si
Quantity 10 g
Synonym 1,1,1-Trifluoromethanesulfonic acid (1,1-dimethylethyl)dimethylsilyl ester, (1,1-Dimethylethyl)dimethylsilyl trifluoromethanesulfonate, Trifluoromethanesulfonic acid (1,1-dimethylethyl)dimethylsilyl ester, Trifluoromethanesulfonic acid tert-butyldimethylsilyl ester, tert-Butyldimethylsilyl triflate, tert-Butyldimethylsilyl trifluoromethanesulfonate, tert-Butyldimethylsilyl trifluoromethanesulfonate, t-Butyldimethylsilyl trifluoromethanesulfonate, TBDMSOTf
InChI Formula InChI=1S/C7H15F3O3SSi/c1-6(2,3)15(4,5)13-14(11,12)7(8,9)10/h1-5H3
SMILES CC(C)(C)[Si](C)(C)OS(=O)(=O)C(F)(F)F
IUPAC Name [tert-butyl(dimethyl)silyl] trifluoromethanesulfonate
Molecular Weight (g/mol) 264.34
Formula Weight 264.0463
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.