missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyldimethylsilylacetylene, 98%

Product Code. 15440378
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15440378 1 g 1g
15450378 5 g 5g
15460378 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15440378 Supplier Thermo Scientific Alfa Aesar Supplier No. H53494.03
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 86318-61-8 | C8H16Si | 140.301 g/mol

tert-Butyldimethylsilylacetylene may be used in the synthesis of β-alkynylketone and β-alkynyl aldehydes. It participates in Cadiot-Chodkiewicz cross-coupling reaction with various bromoalkynes to afford synthetically useful unsymmetrical diynes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
tert-Butyldimethylsilylacetylene may be used in the synthesis of β-alkynylketone and β-alkynyl aldehydes. It participates in Cadiot-Chodkiewicz cross-coupling reaction with various bromoalkynes to afford synthetically useful unsymmetrical diynes.

Solubility
Sparingly soluble in water (0.043 g/L at 25°C).

Notes
Stable under recommended storage conditions.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 86318-61-8
Molecular Formula C8H16Si
Molecular Weight (g/mol) 140.301
MDL Number MFCD00191877
InChI Key RTYNRTUKJVYEIE-UHFFFAOYSA-N
Synonym tert-butyldimethylsilyl acetylene, tert-butyldimethylsilylacetylene, tert-butyl ethynyl dimethylsilane, ethynyl-t-butyldimethylsilane, tert-butyldimethylsilyl ethyne, tert-butyl-ethynyl-dimethyl-silane, silane, 1,1-dimethylethyl ethynyldimethyl, t-butyldimethylsilylacetylene, tert-butyldimethylsilylethyne, tert-butylethynyldimethylsilane
PubChem CID 2757281
IUPAC Name tert-butyl-ethynyl-dimethylsilane
SMILES CC(C)(C)[Si](C)(C)C#C

Specifications

Boiling Point 130°C
Quantity 1 g
Linear Formula (CH3)3CSi(CH3)2C≡CH
UN Number UN1993
Beilstein 1921466
Flash Point 6°C (43°F)
Solubility Information Sparingly soluble in water (0.043g/L at 25°C).
Formula Weight 140.3
Percent Purity 98%
Refractive Index 1.451
Density 0.751
Chemical Name or Material tert-Butyldimethylsilylacetylene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.