missing translation for 'onlineSavingsMsg'
Learn More

tert-Butylhydrazine hydrochloride, 98%, Thermo Scientific Chemicals

Product Code. 10753571 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10753571 500 g Glass bottle 500g
10042290 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10753571 Supplier Thermo Scientific Acros Supplier No. 187085000
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7400-27-3
Molecular Formula C4H13ClN2
Molecular Weight (g/mol) 124.61
MDL Number MFCD00012947
InChI Key DDPWVABNMBRBFI-UHFFFAOYSA-N
Synonym tert-butylhydrazine hydrochloride, hydrazine, 1,1-dimethylethyl-, monohydrochloride, tert-butylhydrazinium chloride, tert-butylhydrazine monohydrochloride, t-butylhydrazine hydrochloride, hydrazine, 1,1-dimethylethyl-, hydrochloride 1:1, 1,1-dimethylethyl hydrazine monohydrochloride, 2-tert-butylhydrazinium chloride, t-butylhydrazine.hcl, t-bunhnh2.hcl
PubChem CID 81889
IUPAC Name tert-butylhydrazine;hydrochloride
SMILES [H+].[Cl-].CC(C)(C)NN

Specifications

Melting Point 190°C to 194°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula (CH3)3CNHNH2·HCl
Quantity 500 g
Beilstein 04, III, 1734
Fieser 12,87
Formula Weight 124.61
Percent Purity 98%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material tert-Butylhydrazine hydrochloride, 98%
Show More Show Less
compliance-icons
Product Identifier
  • tert-Butylhydrazine hydrochloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.