missing translation for 'onlineSavingsMsg'
Learn More

tert-Butylhydrazine hydrochloride, 98%

Product Code. 11426726
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11426726 500 g 500g
11406726 25 g 25g
11416726 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11426726 Supplier Thermo Scientific Alfa Aesar Supplier No. B23876.36
 Restricted Item
Estimated Shipment: 30-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 7400-27-3 | C4H13ClN2 | 124.61 g/mol

It is used in organic synthesis for aromatic nucleophilic N-N exchange reaction. tert-Butylhydrazine Hydrochloride is also used in the preparation of insect growth regulator. tert-Butylhydrazine hydrochloride was used in the synthesis of diphosphinohydrazines, 5-amino-1-tert-butyl-3-(p-chlorophenyl)-4-cyanopyrazole and 1,3,4,5-tetrasubstituted pyrazole derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in organic synthesis for aromatic nucleophilic N-N exchange reaction. tert-Butylhydrazine Hydrochloride is also used in the preparation of insect growth regulator. tert-Butylhydrazine hydrochloride was used in the synthesis of diphosphinohydrazines, 5-amino-1-tert-butyl-3-(p-chlorophenyl)-4-cyanopyrazole and 1,3,4,5-tetrasubstituted pyrazole derivatives.

Solubility
Solubility in water (almost transparency). Soluble in DMSO and methanol.

Notes
Hygroscopic. Store away from water/moisture and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas. Protect from humidity and water.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7400-27-3
Molecular Formula C4H13ClN2
Molecular Weight (g/mol) 124.61
MDL Number MFCD00012947
InChI Key DDPWVABNMBRBFI-UHFFFAOYSA-N
Synonym tert-butylhydrazine hydrochloride, hydrazine, 1,1-dimethylethyl-, monohydrochloride, tert-butylhydrazinium chloride, tert-butylhydrazine monohydrochloride, t-butylhydrazine hydrochloride, hydrazine, 1,1-dimethylethyl-, hydrochloride 1:1, 1,1-dimethylethyl hydrazine monohydrochloride, 2-tert-butylhydrazinium chloride, t-butylhydrazine.hcl, t-bunhnh2.hcl
PubChem CID 81889
SMILES [H+].[Cl-].CC(C)(C)NN

Specifications

Melting Point ∼192°C (decomposition)
Quantity 500 g
Beilstein 3651224
IUPAC Name hydrogen tert-butylhydrazine chloride
Formula Weight 124.61
Percent Purity 98%
Chemical Name or Material tert-Butylhydrazine hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.