missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Tertiapin-Q

Click to view available options
Quantity:
1 mg
Unit Size:
1mg
Description
Tertiapin-Q is a high affinity blocker for inward-rectifier K+ channels. Tertiapin-Q is a stable derivative of the bee venom toxin tertiapin. Tertiapin-Q binds to ROMK1 (Kir1.1) and GIRK1/4 (Kir3.1/3.4) channels with high affinity (Ki values are 1.3 and 13.3 nM respectively) and is selective over Kir2.1 channels. Tertiapin-Q improves heart rate and atrioventricular conduction in a mouse model of bradycardia. Derivative Tertiapin LQ (Cat. No. 4339) also available.
Specifications
Specifications
| Purity | 0.95 |
| Content And Storage | Store at -20°C |
| Description | Tertiapin-Q |
| Molecular Weight (g/mol) | 2452 |
| Product Type | Tertiapin-Q |
| Quantity | 1 mg |
| Sequence | Ala-Leu-Cys-Asn-Cys-Asn-Arg-Ile-Ile-Ile-Pro-His-Gln-Cys-Trp-Lys-Lys-Cys-Gly-Lys-Lys-NH2 (Disulfide bridges: 3 - 14, 5 - 18) |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction