missing translation for 'onlineSavingsMsg'
Learn More

Bunzl Healthcare™ Terumo™ Eccentric Luer-Slip Syringe

Product Code. 12917436
Change view
Click to view available options
Volume (Metric):
2.5 mL
20 mL
50 mL
Graduations:
0.1 mL
1 mL
Unit Size:
Pack of 100
Pack of 25
Pack of 50
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Graduations unitSize
12917436 2.5 mL 0.1 mL Pack of 100
11507302 20 mL 1 mL Pack of 50
13148023 20 mL 1 mL Pack of 50
13114794 50 mL 1 mL Pack of 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 12917436 Supplier Bunzl Healthcare™ Supplier No. TUSS02SE1

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Manufactured with polypropylene for superior thermal and chemical resistance. Bunzl™ Terumo™ Eccentric Luer-Slip Syringes have a unique bevel design to facilitate the penetration of the skin and minimize trauma.

  • Clear polypropylene barrel and plunger; double contact ring
  • Integrated plunger stop prevents accidental pull-out
  • Sterile, Latex-free
  • Bold, precise scale markings
  • Complies to ISO 7886 (DIS 7886)
  • Individually wrapped in tamper evident peel packs
  • Supplied without needles
  • Suitable for gases or liquids
  • Eccentric luer lock
For needles, please reference our disposable needles range, ISO 7886 (DIS 7886): 13127634, 13117614, 13144724, 13187434, 12349219, 13104354.

For research use only, unless otherwise indicated.

Specifications

Certifications/Compliance ISO 7886 (DIS 7886)
Material Polypropylene
For Use With (Application) For research use, for gases or liquids
Product Type Eccentric Luer-slip Syringe
Disposable Single-use
Graduations 0.1 mL
Quantity 100/Pk.
Volume (Metric) 2.5 mL
Connection Type Eccentric Luer Slip
Graduated Yes
Sterility Sterile
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.