Learn More
Terumo™ Hypodermic Needles
Product Code. 13144724
Change view
Click to view available options
Needle Gauge:
21 G
23 G
25 G
Length (English) Needle:
1 in.
Needle Point Style:
Thin Wall
Ultra-thin Wall
Unit Size:
Pack of 100
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Needle Gauge Length (English) Needle Needle Point Style unitSize
13144724 23 G 1 in. Ultra-thin Wall Pack of 100
12810126 21 G 1 in. Ultra-thin Wall -
13117614 25 G 1 in. Thin Wall Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 13144724 Supplier Terumo™ Supplier No. TUAN2325R1
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Provides ultra sharp needles, which translate into greater comfort for the patient.
  • Double bevel design
  • Precision manufactured ultrathin wall
Specifications
Needle Gauge 23 G
Latex Free Yes
Length (English) Needle 1 in.
Product Type Double Bevel Design
Needle Point Style Ultra-thin Wall