missing translation for 'onlineSavingsMsg'
Learn More

TES, 0.2M buffer soln., pH 7.0, Thermo Scientific™

Catalog Number 15435629
Click to view available options
Quantity:
100 mL
250 mL
This item is not returnable. View return policy
70331-82-7
C6H14NNaO6S
251.23
MFCD00065482
GUXMFAHOYNRHBI-UHFFFAOYSA-M
23702152
sodium;2-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]ethanesulfonate
[Na+].OCC(CO)(CO)NCCS([O-])(=O)=O
This item is not returnable. View return policy
70331-82-7
C6H14NNaO6S
251.23
MFCD00065482
GUXMFAHOYNRHBI-UHFFFAOYSA-M
23702152
sodium;2-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]ethanesulfonate
[Na+].OCC(CO)(CO)NCCS([O-])(=O)=O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Concentration 0.2 M
pH 7.0
Physical Form Liquid
Quantity 100 mL
Chemical Name or Material TES buffer soln.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'