missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human TESK1, GST Tag Recombinant Protein

Product Code. 15930313
Click to view available options
Quantity:
1 mg
10 μg
100 μg
Unit Size:
100µg
10µg
1mg
This item is not returnable. View return policy

Product Code. 15930313

Brand: Invitrogen™ A30985

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. This TESK1 recombinant human protein (full length) was expressed in insect cells. TESK1 recombinant human protein is also available in our SelectScreen™ Kinase Profiling Service. Validated for use with the LanthaScreen™ Eu Kinase Binding assay Available in multiple pack sizes for convenient use Also available in our SelectScreen Kinase Profiling Service Protein Form: Full Length, Recombinant.

TESK1 serine/threonine kinase or testis-specific kinase 1 contains an N-terminal protein kinase domain and a C-terminal proline-rich domain which is most closely related to those of the LIM motif-containing protein kinases (LIMKs). TESK1 protein can phosphorylate myelin basic protein and histone in vitro and plays an important role during and after the meiotic phase of spermatogenesis. TESK1 is mainly expressed in testicular germ cells.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q15569
Concentration See Label
For Use With (Application) Kinase Assay
Formulation 50mM tris HCl with 50mM NaCl, 0.25mM DTT, 0.1mM PMSF, 0.1mM EDTA, 10mM glutathione, 25% glycerol and no preservative; pH 7.5
Gene ID (Entrez) 7016
Molecular Weight (g/mol) 93.9 kDa
Name Human TESK1, GST Tag
pH Range 7.5
Purification Method Purified
Quantity 10 μg
Storage Requirements -80°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias AI326901; dual specificity testis-specific protein kinase 1; Tesk1; Testicular protein kinase 1; testis specific protein kinase 1; testis-specific kinase 1
Common Name TESK1
Gene Symbol TESK1
KinaseFamily LISK Kinase Family
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag GST-tag
Sequence Full length
Content And Storage -80°C, Avoid Freeze/Thaw Cycles
Shipping Condition Dry Ice
Expression System Insect cells
Protein Family Kinases & Inhibitors
Protein Form Recombinant, Full Length
Form Liquid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.