missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human TESK1, GST Tag Recombinant Protein
This TESK1 Recombinant Human Protein (full length) was expressed in insect cells.
Brand: Invitrogen™ A32903
Additional Details : Weight : 0.00900kg
Description
TESK1 serine/threonine kinase or testis-specific kinase 1 contains an N-terminal protein kinase domain and a C-terminal proline-rich domain which is most closely related to those of the LIM motif-containing protein kinases (LIMKs). TESK1 protein can phosphorylate myelin basic protein and histone in vitro and plays an important role during and after the meiotic phase of spermatogenesis. TESK1 is mainly expressed in testicular germ cells.- Validated for use with the LanthaScreen™ Eu Kinase Binding assay
- Available in multiple pack sizes for convenient use
Specifications
NP_006276 | |
93.9 kDa | |
LISK Kinase Family | |
Human | |
GST Tag | |
Dry Ice | |
Full Length, Recombinant |
Kinase Assay | |
100 μg | |
Full Length | |
Recombinant | |
Storage Buffer: 50 mM Tris-HCl, pH 7.5, 50 mM NaCl, 10 mM glutathione, 0.1 mM EDTA, 0.25 mM DTT, 0.1 mM PMSF, 25% glycerol | |
Insect | |
Liquid |