missing translation for 'onlineSavingsMsg'
Learn More

Hach™ Test 'N Tube (TNT) Cuvette Tests

Product Code. p-7115597
Click to view available options
For Use With (Application):
Total Phosphate Determination
Unit Size:
50 tests
Each
This item is not returnable. View return policy

Product Code. 12037032

Brand: Hach™ 2742645

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Provides a self-contained package for mixing and measurement. Hach™ Test 'N Tube (TNT) Hach™ Test 'N Tube (TNT) Cuvette Tests include 16mm capped tubes and all necessary reagents in one convenient package for reliable results and easy handling.

Provides a self-contained package for mixing and measurement. Hach™ Test 'N Tube (TNT) Hach™ Test 'N Tube (TNT) Cuvette Tests include 16mm capped tubes and all necessary reagents in one convenient package for reliable results and easy handling.

Test 'N Tube products enable convenient testing with ready to use ampules and pre-measured reagents all packed into one convenient package. Available for determinations of a variety of parameters.

  • Test 'N Tube: Capped 16mm vials provide a safe, self-contained package for mixing and measurement
  • Powder Pillows: Premeasured, single dose packets of powdered reagent
  • Dropper bottles: For easy addition of liquid reagents
  • Precisely dosed reagents and test cuvettes for easy planning and inventory control

Specifications

Description Colorimetric determination cuvette tests
For Use With (Application) Total Phosphate Determination
For Use With (Equipment) Hach DR900
Includes 16mm Tubes, Potassium persulfate Powder Pillows, PhosVer 3 Powder Pillows, Sodiumhydroxyde solution 1.54N, Sulfuric acid solution 1.37N
No. of Tests 25 to 50 (depending on blanks)
Quantity 1 Kit
Range 0.06 to 3.50mg/L PO4
Sufficient For Low Range
Test Method PhosVer 3 w/ Acid Persulfate Digestion
Type Test 'N Tube
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.