Learn More
ITS Europe™ Ultra-High Free Chlorine Test Strips
Description
- Very high range of detection.
- Detection range: 0 - 750ppm (mg/L).
- Designed to resist interference from monochloramines
- Ideal product when testing in a food processing environment.
- Perfect for determining whether or not you have the correct amount of sanitizer present.
- All reagents are impregnated on the test strip pad, and no external chemicals are needed to perform the water quality test.
- Test time: 31 seconds.
Specifications
Specifications
| Analytes | Chlorine |
| No. of Tests | 50 |
| Quantity | 50/Bottle |
| Range | 0 - 750ppm (mg/L) |
Product Title
Haben Sie Verbesserungsvorschläge?