missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Test Tube Racks for MaxQ™ Shaker

Mix and match these shaker test tube racks, available in a variety of sizes and arrays, to suit individual needs in the laboratory.

Brand:  Thermo Scientific™ 30182

Code : 50

Additional Details : Weight : 0.63000kg

 View more versions of this product

Product Code. 10031913

  • 356.00 € / Each
In Stock
to see stock.
Add to basket

Description

Description

Mix and match Thermo Scientific™ MaxQ™ Shaker Test Tube Racks, available in a variety of sizes and arrays, to suit individual needs in the laboratory. All racks are compatible with MaxQ shaker platforms.

Racks are available in a multitude of sizes, arrays, and colors. Compatible with MaxQ shaker platforms.
Specifications

Specifications

Test Tube Rack
6 x 12
14 to 16 mm Tubes, 15 mL Centrifuge Tubes
MaxQ™ Shaker Platform
Orange; 14 to 16 mm; 6 x 12 array (fits 15 mL centrifuge tubes)
Orange
72
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Services