missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Velp Scientifica™ 6 Position Test Tube Stand
Velp Scientifica™ 6 Position Test Tube Stand for use with ECO 6 Thermoreactor.
Brand: Velp Scientifica™ A00001043
Code : A0
Additional Details : Weight : 0.10000kg
Description
- For use with ECO 6 thermoreactor
Specifications
42mm dia. Test Tubes | |
6 Tubes | |
Stand |
Stainless Steel | |
1 Each |