missing translation for 'onlineSavingsMsg'
Learn More

Kartell™ Stopper for Disposable Test Tubes

Product Code. 15305393
Change view
Click to view available options
Color:
Natural
Unit Size:
Pack of 1000
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
15305393 Natural Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15305393 Supplier Kartell™ Supplier No. 8832110
  • ONLINE EXCLUSIVE PRICE
    26.00 €
    24.78 € / Pack of 1000
    Save 1.22 €
    4% Off
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Kartell™ Stopper for Disposable Test Tubes provide a leakproof seal to the test tube, which decreases risk of spilling.

  • CE Marked, IVD Approved
  • Constructed of polyethylene
  • For use with disposable test tubes Kartell™ 88320

Specifications

Certifications/Compliance CE/IVD
Diameter (Metric) Outer 16 mm
Material Polyethylene
Color Natural
For Use With (Equipment) Kartell™ 88320
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.