missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Testosterone-2,2,4,6,6-d5 Benzoate, CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Specifications
Specifications
| Chemical Name or Material | Testosterone-2,2,4,6,6-D5 17-Benzoate |
| Molecular Formula | C26 D5 H27 O3 |
| Quantity | 0.005 g |
| Synonym | (17β)-17-(Benzoyloxy)androst-4-en-3-one-2,2,4,6,6-d5, Testosterone-2,2,4,6,6-D5 17-benzoate, Testosterone-2,2,4,6,6-d5 benzoate, 17β-Hydroxyandrost-4-en-3-one-2,2,4,6,6-d5 benzoate, 17β-Benzoyloxy testosterone-2,2,4,6,6-d5 |
| InChI Formula | InChI=1 S/C26H32O3/c1-25-14-12-19(27)16-18(25)8-9-20-21-10-11-23(26(21,2)15-13-22(20)25)29-24(28)17-6-4-3-5-7-17/h3-7,16,20-23 H,8-15H2,1-2H3/t20-,21-,22-,23-,25-,26-/m0/s1/i8D2,12D2,16 D |
| SMILES | [2 H]C1=C2C([2 H])([2 H])C[C@H]3 [C@@H]4 CC[C@H](OC(=O)c5ccccc5)[C@@]4(C)CC[C@@H]3 [C@@]2(C)CC([2 H])([2 H])C1=O |
| IUPAC Name | [(8 R,9 S,10 R,13 S,14 S,17 S)-2,2,4,6,6-pentadeuterio-10,13-dimethyl-3-oxo-1,7,8,9,11,12,14,15,16,17-decahydrocyclopenta[a]phenanthren-17-yl] benzoate |
| Molecular Weight (g/mol) | 397.561 |
| Formula Weight | 397.267 g/mol |
| Percent Purity | 99 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction