missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Testosterone Parameter Assay Kit

Product Code. 18731844
Click to view available options
Sufficient For:
1 x 96-well Plate
6 x 96-well Plates
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 18731844

Brand: R&D Systems™ SKGE010

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Testosterone Parameter Assay Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Testosterone Parameter Assay Kit quantifies multi-species Testosterone in cell culture supernates (100 ul), serum (20 ul), edta plasma (20 ul), heparin plasma (20 ul).
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 0.04 - 10ng/mL
Assay Sensitivity 0.041ng/mL
Conjugate Biotin
Cross Reactivity Cross-species reactivity not tested. Cross-reactivity observed with 1 or more available related molecules.
Product Type ELISA Kit
Sample Type Cell Culture Supernatant, Plasma (EDTA), Plasma (Heparin), Serum
Specificity Testosterone Levels in Various Samples
Sufficient For 6 x 96-well Plates
Target Species Multi-species
For Use With (Application) ELISA
Quantity 1 Kit
Sample Volume Cell Culture Supernates (100uL),Serum (20uL),EDTA Plasma (20uL),Heparin Plasma (20uL)
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.