missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Testosterone Parameter Assay Kit
Click to view available options
Sufficient For:
1 x 96-well Plate
6 x 96-well Plates
Unit Size:
1 set
Description
The Testosterone Parameter Assay Kit quantifies multi-species Testosterone in cell culture supernates (100 ul), serum (20 ul), edta plasma (20 ul), heparin plasma (20 ul).
Specifications
Specifications
| Assay Range | 0.04 - 10ng/mL |
| Assay Sensitivity | 0.041ng/mL |
| Conjugate | Biotin |
| Cross Reactivity | Cross-species reactivity not tested. Cross-reactivity observed with 1 or more available related molecules. |
| Product Type | ELISA Kit |
| Sample Type | Cell Culture Supernatant, Plasma (EDTA), Plasma (Heparin), Serum |
| Specificity | Testosterone Levels in Various Samples |
| Sufficient For | 1 x 96-well Plate |
| Target Species | Multi-species |
| For Use With (Application) | ELISA |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction