missing translation for 'onlineSavingsMsg'
Learn More

Testosterone Undecanoate, TRC

Product Code. 30409628
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30409628 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30409628 Supplier Toronto Research Chemicals Supplier No. TRCT155050100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Testosterone Undecanoate
CAS 5949-44-0
Molecular Formula C30 H48 O3
Quantity 100 mg
Synonym Androst-4-en-3-one','17-[(1-oxoundecyl)oxy]-','(17β)-, Testosterone','undecanoate (6CI,7CI,8CI), Undecanoic acid','ester with testosterone (8CI), Andriol, Andriol Testocaps, Nebido, Org 538, Testocaps, Testosterone undecylate, Undestor
InChI Formula InChI=1S/C30H48O3/c1-4-5-6-7-8-9-10-11-12-28(32)33-27-16-15-25-24-14-13-22-21-23(31)17-19-29(22,2)26(24)18-20-30(25,27)3/h21,24-27H,4-20H2,1-3H3/t24-,25-,26-,27-,29-,30-/m0/s1
SMILES CCCCCCCCCCC(=O)O[C@H]1CC[C@H]2[C@@H]3CCC4=CC(=O)CC[C@]4(C)[C@H]3CC[C@]12C
IUPAC Name [(8R,9S,10R,13S,14S,17S)-10,13-dimethyl-3-oxo-1,2,6,7,8,9,11,12,14,15,16,17-dodecahydrocyclopenta[a]phenanthren-17-yl] undecanoate
Molecular Weight (g/mol) 456.7
Formula Weight 456.3603
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.