Learn More
Invitrogen™ Tetra Buffer Dam
Description
The Invitrogen Tetra Buffer Dam is designed for use with the Tetra Electrophoresis primary (Cat. No. 1658037) and companion (Cat. No. 1658038) cores to provide flexibility in the number of gels run in Bio-Rad's Mini-PROTEAN Tetra Cell. This buffer dam fits into the empty gel slot, creating a barrier between the cathode and anode chambers, ensuring optimal performance and efficiency for your electrophoresis runs.
- Enable gel flexibility—allows for flexible gel running by fitting into the empty gel slot
- Maintain performance—creates a barrier between cathode and anode chambers to ensure optimal electrophoresis conditions
- Integrate seamlessly—compatible with Tetra Electrophoresis primary and companion cores for easy adoption
- Maximal compatibility—compatible with other Invitrogen mini tanks such as the Mini Gel Tank (Cat. No. A25977) and the XCell SureLock Mini-Cell (Cat. No. EI0001)
The Tetra Buffer Dam is essential for labs needing flexibility in the number of gels run per core, as it maintains the integrity of the electrophoresis setup by occupying the empty slot. This ensures that the electrophoresis conditions remain consistent, leading to reliable and reproducible results. The buffer dam is compatible with both the Tetra Electrophoresis primary and companion cores, making it a versatile addition to your existing electrophoresis equipment.
Using the Tetra Buffer Dam is straightforward: simply insert it into the empty gel slot in the Tetra primary or companion core when running fewer gels. This creates a barrier that maintains the separation between the cathode and anode chambers, ensuring that your electrophoresis runs are efficient and effective. By using the buffer dam, you can achieve consistent results without the need for additional equipment or complex adjustments.
The Tetra Buffer Dam is an economical solution that enhances the flexibility and efficiency of your electrophoresis setup. It ensures that you can run a variable number of gels without compromising on performance, making it an ideal accessory for any lab using Bio-Rad's Mini-PROTEAN Tetra Cell with Invitrogen precast protein gels.
Specifications
Specifications
| For Use With (Equipment) | Bio-Rad Mini-PROTEAN Tetra Cell |
| Type | Buffer Dam |
| Quantity | 1 Ea. |
For Research Use Only.
Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.