missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Tetra Core Set and Bolt™ Bis-Tris Plus 4-12% Gels Combos

Product Code. 30484925
Change view
Click to view available options
Wells:
10-well
12-well
15-well
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Wells unitSize
30484925 10-well Each
30485053 12-well Each
30484887 15-well Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30484925 Supplier Invitrogen™ Supplier No. NW0412TS10W
  • ONLINE EXCLUSIVE PRICE
    540.00 €
    437.40 € / Pack of 30
    Save 102.60 €
    19% Off
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

The Tetra Electrophoresis Core Set and Bolt Bis-Tris Plus 4-12% Combo includes the Tetra Electrophoresis primary and companion cores along with three boxes of Bolt Bis-Tris Plus gels. Easily swap and run up to four Bolt Bis-Tris gels in Bio-Rad's Mini-PROTEAN™ Tetra Cell tank with a run time as short as 20 minutes.

The Invitrogen Tetra Electrophoresis Core Set and Bolt Bis-Tris Plus 4-12% Combo provides a comprehensive solution for running high-performance Invitrogen precast protein gels in Bio-Rad's Mini-PROTEAN Tetra Cell. This combo product includes the Tetra Electrophoresis primary and companion cores along with three boxes of Bolt Bis-Tris Plus gels, allowing for seamless integration and efficient protein analysis applications such as western blotting and SDS-PAGE. Its intuitive design ensures ease of use, making it accessible for both new and experienced users.

Contents of the combo kit

  • Tetra Electrophoresis Primary Core (Cat. No. 1658037)
  • Tetra Electrophoresis Companion Core (Cat. No. 1658038)
  • 3 boxes of Bolt Bis-Tris Plus 4-12% gels (10 gels per box)

About the Tetra Electrophoresis Core Set

The Tetra Electrophoresis cores stand out due to their compatibility with all Invitrogen mini gels, allowing users to swap and run gels effortlessly while achieving high-performance results. When using both the primary and companion cores, up to four gels can be run simultaneously, which significantly increases throughput and efficiency. Additionally, the self-aligning cassettes and leak-proof design ensure stable and reliable electrophoresis runs, making experiment setup easy and fast.

The Tetra Electrophoresis Primary Core comes with the Tetra Buffer Dam (Cat. No. 1658039) for use when running only one gel. To run three or four gels at once, the Tetra Electrophoresis Companion Core is used alongside the Primary Core. This combination allows for the simultaneous running of multiple gels, further enhancing throughput and efficiency in your lab. The Companion Core works seamlessly with the Primary Core, providing a flexible solution for varying experimental needs.

This cost-effective solution eliminates the need for new electrophoresis equipment, saving costs while enhancing your lab's capabilities. The Tetra Electrophoresis Core Set is straightforward to use: simply swap the cores in your Bio-Rad Mini-PROTEAN Tetra system for the Invitrogen Tetra cores and use Invitrogen mini protein gels following their recommended protocols.

About Bolt Bis-Tris Plus Gels

Bolt Bis-Tris Plus gels are precast polyacrylamide gels designed for optimal broad range separation of proteins under denaturing conditions. Similar to NuPAGE Bis-Tris gels, Bolt Bis-Tris Plus gels offer excellent protein integrity with neutral pH, a wide range of molecular weight separation, fast run times of 20–35 minutes, and a long shelf life of up to 16 months at room temperature. Bolt Bis-Tris Plus gels also have the unique WedgeWell design that allows for easier sample loading and up to 2X more sample volume than other standard precast gels. Bolt Bis-Tris Plus gels come in four percentages and multiple well formats. Each box contains 10 gels.

TRUSTED_SUSTAINABILITY

Specifications

Adapters Tetra Electrophoresis Cores
Content And Storage Store at 4°C to 25°C. Do not freeze.
Buffer Bolt MES SDS Running Buffer, Bolt MOPS SDS Running Buffer
Gel Percentage 4 to 12%
Gel Size Mini
Gel Thickness 1.0 mm
Gel Type Bis-Tris Plus
Sample Loading Volume Up to 60 μL
Separation of Protein
Separation Range 3.5 to 260 kDa
Separation Type Denaturing
Wells 10-well
Mode of Separation Molecular Weight
Quantity Tetra Core Set, 3 boxes of gels (30 gels)
Width (Metric) 8 cm
Product Line Bolt, WedgeWell
Product Type Electrophoresis Cores and Precast Protein Gels
For Use With (Equipment) Bio-Rad Mini Protean Tanks
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.