missing translation for 'onlineSavingsMsg'
Learn More

Tetra-n-butylammonium borohydride, 97%

Product Code. 11422000
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11422000 250 g 250g
11402000 10 g 10g
11412000 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11422000 Supplier Thermo Scientific Alfa Aesar Supplier No. A17494.30
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 33725-74-5 | C16H40BN | 257.31 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 33725-74-5
Molecular Formula C16H40BN
Molecular Weight (g/mol) 257.31
MDL Number MFCD00012035
InChI Key GMBOFJFPOCGSOI-UHFFFAOYSA-N
Synonym tetrabutylammonium borohydride, tetra-n-butylammonium borohydride, n,n,n-tributylbutan-1-aminium tetrahydroborate, boron 1-; tetrabutylazanium, boron 1-; tetrabutylammonium
PubChem CID 9881569
SMILES [BH4-].CCCC[N+](CCCC)(CCCC)CCCC

Specifications

Melting Point 125°C to 131°C
Flash Point 140°C (284°F)
Linear Formula [CH3(CH2)3]4NBH4
Quantity 250 g
UN Number UN3131
Sensitivity Moisture sensitive
IUPAC Name boron(1-);tetrabutylazanium
Formula Weight 257.31
Percent Purity 98%
Chemical Name or Material Tetra-n-butylammonium borohydride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.