missing translation for 'onlineSavingsMsg'
Learn More

Tetra-n-butylammonium bromide, 98+%

Product Code. 11483953
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11483953 2500 g 2500g
11463953 100 g 100g
11473953 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11483953 Supplier Thermo Scientific Alfa Aesar Supplier No. A10249.0E
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 1643-19-2 | C16H36BrN | 322.375 g/mol

Phase transfer catalyst Tetra-n-butylammonium bromide is used as a phase transfer catalyst for the preparation of lactones by cyclization of potassium salt of μ-bromocarboxylic acids and beta-lactones from beta-bromoacid by cyclization. It is also involved in the Curtius rearrangement, N-alkylation of aromatic carboxamides. It plays an important role as an ionic liquid in the regioselective O-alkylation reactions. It is also used in the preparation of polymer solar cells and single component green-light emitting electrochemical cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phase transfer catalyst Tetra-n-butylammonium bromide is used as a phase transfer catalyst for the preparation of lactones by cyclization of potassium salt of µ-bromocarboxylic acids and beta-lactones from beta-bromoacid by cyclization. It is also involved in the Curtius rearrangement, N-alkylation of aromatic carboxamides. It plays an important role as an ionic liquid in the regioselective O-alkylation reactions. It is also used in the preparation of polymer solar cells and single component green-light emitting electrochemical cells.

Solubility
Soluble in water and ethanol.

Notes
Hygroscopic. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1643-19-2
Molecular Formula C16H36BrN
Molecular Weight (g/mol) 322.375
MDL Number MFCD00011633
InChI Key JRMUNVKIHCOMHV-UHFFFAOYSA-M
Synonym tetrabutylammonium bromide, tetra-n-butylammonium bromide, tbab, tetrabutylazanium bromide, tetrabutyl ammonium bromide, tetrabutyl-ammonium bromide, aliquat 100, tbabr, ipc-tba-br, unii-vjz168i98r
PubChem CID 74236
ChEBI CHEBI:51993
IUPAC Name tetrabutylazanium;bromide
SMILES CCCC[N+](CCCC)(CCCC)CCCC.[Br-]

Specifications

Melting Point 101°C to 105°C
Density 1.15
Odor Amine-like
Linear Formula [CH3(CH2)3]4NBr
Quantity 2500 g
Beilstein 3570983
Sensitivity Hygroscopic
Solubility Information Soluble in water and ethanol.
Formula Weight 322.38
Percent Purity ≥98%
Chemical Name or Material Tetra-n-butylammonium bromide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.